Goggles, Vistamax VNC21
Catalog # DALL1002759
Supplier: HONEYWELL SAFETY
Specifications
- Type:Anti-fog, anti-scratch, chemical resistant
- Colour:Red
- Lenses:Clear Acetate/PC
- Pk:1
Specifications
About this item
High performance anti-fog goggles, suitable for most very humid environments.
- No vent frame gives excellent protection againsts large and fine dust particles as well as droplets of liquids
- Unique anti-fog twin lens system: Front acetate lens for chemical resistance, rear polycarbonate lens for impact resistance
- Thermal bridge with aluminimum condenser
Designed for petrochemical, industrial or laboratory, administration or foundry.
Certifications: ISO 9001/2000