Cleanroom bond papers, Munising LP®
Munising LP® bond papers are saturated with synthetic polymers and are free of inorganic fillers which prevents the doping of products and accepts standard printing inks without skipping, smudging or smearing.
- Natural latex free
- Heat resistant for use in laser printers and photocopy machines
- Autoclavable