Order Entry
Finland
ContactUsLinkComponent
Accessories for face shields
Accessories for face shields
Catalog # UNVT605.04.00.00
Supplier:  UNIVET
undefined
Accessories for face shields
Catalog # UNVT605.04.00.00
Supplier:  UNIVET
Specifications
  • Description:
    Ergonomic headband with foam sweatband and 90° flip-up function, protects against electric arcs, blue, for head sizes 53 - 61 cm
  • Pk:
    1
Specifications
About this item
This ergonomic headgear is made from anti-allergic material. With upper band for height adjustment of the cap.
  • Interchangeable front foam sweatband
  • Adjustment of the head circumference by rack
  • 90° flip-up through friction
  • Free of exposed metal parts for protection against electric arcs