Order Entry
Finland
ContactUsLinkComponent
Face shield, medical, 710
Face shield, medical, 710
Catalog # UNVT710.00.00.00
Supplier:  UNIVET
undefined
Face shield, medical, 710
Catalog # UNVT710.00.00.00
Supplier:  UNIVET
Specifications
  • Type:
    Anti-fog
  • Description:
    Face shield with clear lens and blue frame
  • Pk:
    1
Specifications
About this item
This face shield features an adjustable elastic band and an anallergic, interchangeable foam at the front.
  • Interchangeable
  • 90° flip up
Weight: 97 g
EN 166 (3 S CE)