Vinyl gloves, Evercare® Vinyl Soft
Supplier: Onemed Sverige AB
Certificates
About this item
Powder-free and powdered evercare® vinyl Soft gloves offer protection for many applications. The 100% synthetic glove is somewhat softer than the standard vinyl and has a good feel and performance.
- DOP-free
- No latex proteins, good alternative for those suffering from Type I allergies
- Suitable for food handling, except for fatty food
- Very soft
Certifications: EN 455; AQL 1,5
Food contact approved
Food contact approved
Specifications
- Catalog No:
- SELA6302
- SELA6303
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Latex / latex-free:
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powder-free
- Powder-free
- Cuff style:
- Beaded
- Beaded
- EN norm:
- EN 455
- EN 455