Order Entry
Finland
ContactUsLinkComponent
Analogue caliper
Analogue caliper
  MOIN1452MI
 :  MOINEAU
undefined
Analogue caliper
  MOIN1452MI
 :  MOINEAU

 

  • Range:
    0 - 150 mm
  • Resolution:
    1:50
  • Description:
    Analogue caliper
  • Pk:
    1

 

 

Analogue caliper with clamping screw.

  • Depth gauge
  • Clamping screw

Weight: 270 g