Strong cation exchange columns, MAbPac™ SCX-10 RS
Supplier: Thermo Fisher Scientific
Certificates
About this item
BioRS, strong cation exchange columns designed for monoclonal antibodies (MAbs) and associated variants. The unique non porous pellicular resin provides the resolving power to separate MAb variants that differ by as little as one charged residue. A hydrophilic layer surrounds the polymeric beads and eliminates hydrophobic interactions with the resin, resulting in very efficient peaks. A proprietary grafted cation-exchange surface provides pH selectivity control for high-resolution separations.
- UHPLC, high throughput analysis
- Specially developed bio-inert PEEK lined stainless steel column hardware
- High pressure compatibility
Specifications
- Catalog No:
- 555-7373
- 555-7370
- 555-7362
- 555-7375
- 555-7338
- Max. pressure:
- 7000 psi
- 7000 psi
- 7000 psi
- 7000 psi
- 7000 psi
- pH range:
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- Temperature limit:
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C