Sample bags with closure, SteriBag StandUp
Supplier: BURKLE
Certificates
About this item
SteriBag StandUp are sterile sample bags with a stand-up base. The special bottom unfolds as soon as the bag is filled. This makes SteriBag StandUp stable and able to stand on its own without a separate stand or support, leaving both your hands free to take the sample and fill the bag. The wire bracket at the closure keeps the bag open, making it easy and convenient to fill. Can be used for samples available as liquids, powders, solids and pastes; suitable for foods and can be frozen.
- Space-saving, unbreakable, inexpensive and sterile replacement for bottles and beakers
- Easy and safe to transport
- SteriBag StandUp, the sample bag for
- Easy handling of samples
- Stand stably
- Protected against discharge
- Sterilised
- With labelling field