Gel permeation chromatography columns, PLgel individual pore size
Supplier: VARIAN
Certificates
About this item
Individual pore size GPC columns offer high resolution over a specific molecular weight range. The linear portion of the calibration curve, where the slope is at its shallowest, defines the MW region over which optimum resolution will be achieved.
- Very high efficiency
- Fast analysis