Semi-micro and Analytical Balances, BM Series
A highly specified range of analytical balances from A&D with an upper neutralising chamber that houses a built-in fanless ioniser to eliminate static charge on the sample or sample container. Applications include counting with Automatic Counting Accuracy Improvement, percentage weighing, below balance weighing and density determination*. The balances are supplied with a data logger that records up to 5000 results with time and date providing data not only about the weighing operation but also environmental readings from built-in sensors.
- Easy, fanless static elimination using a built-in ioniser
- Interlockable sliding doors allowing efficient operation with both hands
- Twelve weighing units (including one custom unit), overload protection
- Data memory function for 100 weighing results with time and date
- Built-in temperature, humidity, and atmospheric pressure sensors
- Automatic calibration with an internal mass and automatic response adjustment
GLP/GMP/GCP/ISO compliant output.
*Available on models BM-252/200/300/500 with optional density determination kit.
: Please contact your local Avantor sales office for details on other accessories including LAN-Ethernet interface with WinCT-Plus software, density determination kits, analytical pans, remote display and calibration weights.
: Balance supplied with dust plate, support rings and pan, draught shield ring, WinCT (CD-ROM), weighing data logger (AD-1688), tweezers for calibration mass (AD-1689), AC adapter, USB cable, display cover and instruction manual.
: Please note these products are not available in Republic of Ireland or Northern Ireland.