Cleanroom Tables, Heavy Duty
Supplier: Palbam
New Product
Certificates
About this item
These cleanroom heavy duty tables are designed for more robust and heavy duty activities and are manufactured to the highest cleanroom standards. The bases are fabricated from tubing for easy cleaning and optimal airflow.
- The tables have fixtures enabling all the accessories to be attached at time of purchase or in the future if the need arises
- The tops are designed to reduce vibrations and are very strong holding up to 100 kg
Stainless steel 304 with electropolished finish and anodised aluminium connectors.
Delivery: Typically available from stock for direct shipment from the supplier. The tables are shipped flat-packed with easy assembly instructions (assembly time ~10 minutes). Other shelves are available upon request.