Knitted gloves, BO600 / BO605
: Showa
These seamless knitted nylon gloves with elasticated cuffs feature a PU coating on the fingertips. The coating protects the hand from oils and micro cuts.
- Lightweight, elastic, low linting
- Anatomical shape, resistant to deformation
- No fingerprints left on handled objects
- Minimal allergy risk, no irritation by seams
- Available in different lengths: Long version provides protection for the wrist
Ideal for light assembly tasks in the automotive and metalworking industry as well as in logistics.
Colour: White
EN 388 (0131); Cat. II
: EN 388:2016 (0131X); Cat. II.
- Catalog No:
- 112-4311
- 112-4310
- 112-4222
- 112-4309
- 112-4312
- 112-4223
- 112-4220
- 112-4221
- 112-4219
- Model:
- BO600
- BO600
- BO605
- BO600
- BO600
- BO605
- BO605
- BO605
- BO605
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (0131X)
- EN 388:2016 (0131X)
- EN 388:2016 (0131X)
- EN 388:2016 (0131X)
- EN 388:2016 (0131X)
- EN 388:2016 (0131X)
- EN 388:2016 (0131X)
- EN 388:2016 (0131X)
- EN 388:2016 (0131X)