GC septa, Marathon
Supplier: Thermo Fisher Scientific
Certificates
About this item
Made of low-bleed silicone, these septa have excellent mechanical properties and are ideal for demanding GC-MS applications.
- Pre-pierced for reliable performance
- Op to 400 injections per septa
- Maximum temperature 400 °C