Nitrile gloves, Peha-soft® Nitrile Fino
Toimittaja: HARTMANN PAUL
Sertifikaatit
Tietoja tästä tuotteesta
These disposable gloves are made from very soft nitrile. Ideal for use in all areas with contamination risks.
- Inner coating for easier donning
- Tear-resistant, excellent fit and dexterity
- Textured fingertips for improved grip
- Latex-free and therefore ideal for people with type I allergies
Suitable for applications involving the handling of chemicals and disinfectants.
Certifications: EN 455, EN 374; Cat. I+II
Food contact approved
Food contact approved
Tekniset tiedot
- Catalog No:
- HART9421984
- HART9421974
- HART9421994
- Model:
- Nitrile Fino
- Nitrile Fino
- Nitrile Fino
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Finish:
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Cuff style:
- Beaded
- Beaded
- Beaded
- EN norm:
- EN 455, EN 374
- EN 455, EN 374
- EN 455, EN 374
- Food contact approved:
- +
- +
- +