Capillary columns, TraceGOLD™ TG-ALC Plus I and TG-ALC Plus II
Supplier: Thermo Fisher Scientific
Certificates
About this item
Application-specific columns for blood alcohol analysis.
- Low bleed for excellent signal-to-noise ratio, sensitivity and mass spectral integrity
- Fast analysis time giving full separation of blood alcohols
Frequently Bought Together