Capillary columns, TraceGOLD™ TG-624SilMS
Supplier: Thermo Fisher Scientific
Certificates
About this item
High thermal stability column ideal for volatile organic analysis.
- High thermal stability - maximum temperatures up to 320 °C
- Highly inert - excellent peak shape for a wide range of compounds
Limited Time Sale
Select items below are currently on sale
Frequently Bought Together