Capillary columns, TraceGOLD™ TG-5SilMS Fast
These columns are designed for low bleed and reduced susceptibility to oxidation.
- Low polarity, silarylene phase
- High thermal stability - maximum temperatures up to 330/350 °C
Frequently Bought Together
These columns are designed for low bleed and reduced susceptibility to oxidation.