Examination gloves, Peha-soft®
Supplier: HARTMANN PAUL
Certificates
About this item
These disposable examination gloves made of soft latex are ideal for wearers who experience hypersensitivity reactions to glove powder. Also suitable for use in all areas where staff are at particular risk of contamination and need high touch sensitivity and a good fit. May be used in direct contact with food.
- Non sterile, powder-free
- Highly elastic and tear-resistant
- Texturing of the finger area ensures optimum grip
- Double-chlorination for excellent donning, fit and high tactile sensitivity
Certifications: Food contact approved
Specifications
- Catalog No:
- HART9421605
- HART9421615
- HART9421625
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Finish:
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Cuff style:
- Beaded
- Beaded
- Beaded
- Food contact approved:
- +
- +
- +