Sample loops for Cheminert® valves, stainless steel
These VICI Jour sample loops are made of high quality stainless steel 316.
- Fit all Cheminert® C1, C2, C2V, and C3 type valves
- High pressure resistance, low permeability
- Tolerance of ±0,05 mm and pressure rating of < 350 bar
They are cut using an electrolytic cutting process, cleaned with ultrasound and high pressure steam to remove contaminants, dried, and capped with softener-free caps to avoid recontamination of the tubing.
: Supplied with stainless hex-head nuts and stainless ferrules for reliable connections.