ESD safety shoes, lace-up, X-Light, 7131033/7131038
Supplier: Abeba
Certificates
About this item
ESD safety shoe made from smooth leather, reflective inserts, and a PU sole which is resistant to fuels.
- Perspiration-absorbent, Silver Point lining
- Replaceable active comfort insole provides optimised comfort; orthopaedic insoles possible
- Sanitized® treatment reduces odour and bacterial growth
- Lightweight PU sole with high slip resistance and shock absorbing system
- Protective steel toecap (200 J)
Colour: White/black
EN 61340 (ESD), EN ISO 20345:2011; S2, SRA
Certifications: EN 61340 (ESD); DGUV 112-191; EN ISO 20345:2011; S2, SRC.