Order Entry
Finland
ContactUsLinkComponent
Wipes and bottle dispenser
Wipes and bottle dispenser
Catalog # 113-8176
Supplier:  Palbam
undefined
Wipes and bottle dispenser
Catalog # 113-8176
Supplier:  Palbam
Specifications
  • W×D×H:
    395×115×310 mm
  • Pk:
    1
Specifications
About this item
This wall-mounted dispenser is designed for wipes and IPA bottles.
  • No lid, allows one handed use
  • Two holes for stable mounting to wall
  • Stainless steel 304, electropolished finish
Used in industries such as pharmaceutical, biotechnology, medical device, microelectronics and nanotechnology - Essential to cleanroom wipe-down procedures.
Certifications: ISO certified
Delivery: Typically available from stock for direct shipment from the supplier.