Order Entry
Finland
ContactUsLinkComponent
Gown and hood dispenser
Catalog # 113-8175
Supplier:  Palbam
undefined
Gown and hood dispenser
Catalog # 113-8175
Supplier:  Palbam
Specifications
  • Dispenser type:
    Wall-mountable
  • Material:
    Stainless steel
  • PPE type:
    Garments
  • Colour:
    Grey
  • Description:
    Gown and hood dispenser, stainless steel
  • W×D×H:
    300×240×800 mm
  • Pk:
    1
Specifications
About this item
Stainless steel 304, electropolished finish wall-mounted dispenser for plastic wrapped gowns and hoods.
  • Easy refilling and easy retrieval
  • Designed to prevent spillage of items
  • Can be adapted for use as a general dispenser
For use in cleanrooms.
Certifications: ISO certified
Delivery: Typically available from stock for direct shipment from the supplier.