Cell culture media, Glasgow MEM (GMEM BHK 21)
Supplier: BIOWEST
Certificates
About this item
Glasgow MEM (GMEM) was originally formulated for growing BHK 21 cells. The original formulation is an Eagle's Basal Medium modified to contain two times the normal concentration of amino acids and vitamins. It is intended for use with adherent cell lines.