Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Furniture
Storage Cabinets
Accessories,safety underbench cabinets, type 90, uts line
Accessories,safety underbench cabinets, type 90, uts line
Accessories,safety underbench cabinets, type 90, uts line
Supplier:
DUPERTHAL
Certificates
Expand image
Accessories,safety underbench cabinets, type 90, uts line
Supplier:
DUPERTHAL
About this item