Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Pipette Tip Boxes and Racks
Racks for Standard/Eppendorf® pipette tips (bulk)
Racks for Standard/Eppendorf® pipette tips (bulk)
Racks for Standard/Eppendorf® pipette tips (bulk)
Catalog # GREI973202BE
Supplier:
Greiner Bio-One
Expand image
Racks for Standard/Eppendorf® pipette tips (bulk)
Catalog # GREI973202BE
Supplier:
Greiner Bio-One
Specifications
Colour code:
Yellow
Description:
PP rack for bulk tips (100/200 µl), suitable for Eppendorf®
Pk:
50
Specifications
About this item
Product Details
Product Details