Screw caps for tubes
: DELTALAB
Screw caps made of polypropylene and aluminium with an internal liner made of rubber.
- Both models of caps are fully autoclavable and are pharmaceutical quality
- Excellent chemical resistance
- Excellent temperature resistance
Screw caps for round bottom borosilicate glass tubes and flat bottom soda glass tubes.