Tape dispenser
Catalog # LIMP661-008
Supplier: PREMIER LIMPET
Specifications
- For Use With:Tapes with 25 mm width
- Description:Tape dispeneser, 1 position
- Pk:1
Specifications
About this item
Designed for heavy duty service.
- Mounted on non-slip rubber pads