Jars, water sampling, wide mouth, square
Supplier: DELTALAB
Certificates
About this item
PET, transparent, with red PP screw cap.
- Available with or without thiosulphate sodium
- Sterilised by radiation
Wide mouth and grips at two sides, easing the sample collection. Cap with internal liner made of polexan with a label that performs as tamper evident. Each bottle is labelled specifying description, code, lot, and expiry date.
With liquid sodium thiosulphate (24 mg/l): Ideal to analyse water meant for human consumption, swimming pools, and any other water where chlorine may modify the composition of the sample while being transported.
Empty, without thiosulphate: Ideal to sample water for physico-chemical analysis, as well as for other sampling requiring sterile bottles.