Transparent classic human skull model
BINH1000051
- Weight:0,6 kg
- Description:Transparent classic human skull model, in three parts
- W×D×H:200×135×155 mm
- Pk:1
Use this unique transparent replica of the human skull to study internal structures that otherwise are visible only through x-ray images.
- High-quality original casts
- Skull is handmade of hard, unbreakable plastic
- Highly accurate representation of the fissures, foramina, processes, sutures etc.
- Can be disassembled into Skull Cap, Base of Skull and Mandible
- As an option, you can insert a five-part brain (C 18) into the skull