Order Entry
Finland
ContactUsLinkComponent
Micro spoon, Wironit steel
Micro spoon, Wironit steel
Catalog # 231-1410
Supplier:  HAMMACHER KARL
undefined
Micro spoon, Wironit steel
Catalog # 231-1410
Supplier:  HAMMACHER KARL

Specifications

  • Description:
    Micro spoon, single end
  • Length:
    140 mm
  • Seq:
    01
  • Pk:
    1
  • Spoon size:
    5 mm

Specifications

About this item

Single-ended micro spoon made from WIRONIT steel.