Order Entry
Finland
ContactUsLinkComponent
Policemen, rubber
Policemen, rubber
Catalog # HECH41596010
undefined
Policemen, rubber
Catalog # HECH41596010

Specifications

  • Description:
    Rubber policeman, 35×25×32 mm (L×W×H mm)
  • Pk:
    1

Specifications

About this item

Spade shaped wiper made from natural rubber.

Int. Ø: 5 mm.

Ordering information: Supplied without glass rod.