Order Entry
Finland
Orders LinkContactUsLinkComponent
 

Phloretin

Supplier:   Cayman Chemical
Phloretin
Supplier:  Cayman Chemical
Certificates

3 Options Available Below

Product Details & Documents
  • CAS Number:60-82-2
  • Formula:C₁₅H₁₄O₅
  • MDL Number:MFCD00002288
  • Molecular Weight:274.28 g/mol
  • EINECS:200-488-7
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 59,00 €
    59,00 €
    1 * 250 mg
       59,00 €

  • Glass bottle for solids
    500 mg
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 110,00 €
    110,00 €
    1 * 500 mg
       110,00 €

  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 172,00 €
    172,00 €
    1 * 1 g
       172,00 €