Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Acridine orange
Acridine orange
Acridine orange
Acridine orange
Supplier:
Cayman Chemical
Certificates
Expand image
Acridine orange
Supplier:
Cayman Chemical
About this item
CAS Number:
494-38-2
:
Formula:
C₁₇H₁₉N₃
MDL Number:
MFCD00005029
:
265.35 g/mol