
Benzo-18-crown-6 (benzo-1,4,7,10,13,16-hexoxacyclooctadecane) ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:14098-24-9
:
Formula:C₁₆H₂₄O₆
MDL Number:MFCD00062741
:312.36 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 96.0 %
- Melting point:43.0 to 50.0 deg-C
- Solubility in Methanol:very faint turbidity