Order Entry
Finland
ContactUsLinkComponent
 
 
1-Butyl-2,3-dimethylimidazoliumbis(trifluoromethylsulphonyl)imide ≥98.0% (by titrimetric analysis)
 :  TCI
 
undefined
 
1-Butyl-2,3-dimethylimidazoliumbis(trifluoromethylsulphonyl)imide ≥98.0% (by titrimetric analysis)
 :  TCI

 

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Purity (Ion exchange titration):
    min. 98.0 %