
2-Phenylpiperidine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:3466-80-6
:
Formula:C₁₁H₁₅N
MDL Number:MFCD00270125
:161.25 g/mol
Boiling Pt:112°C
EINECS:222-422-6
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %