Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Bisacrylamide
Bisacrylamide 2% in aqueous solution
Bisacrylamide 2% in aqueous solution
Bisacrylamide 2% in aqueous solution
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Bisacrylamide 2% in aqueous solution
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
110-26-9
:
Formula:
(H₂C=CHCONH)₂CH₂
MDL Number:
MFCD00008625
:
154.17 g/mol
Storage Temperature:
Refrigerator
EINECS:
203-750-9
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAJ63265
Videos
Thermo Scientific Acroseal 25 Years
Documentation