Order Entry
Finland
ContactUsLinkComponent
Malachite green oxalate ≥95.0% (by titrimetric analysis) sensitive spectrophotometric reagent for PO₄
Malachite green oxalate ≥95.0% (by titrimetric analysis) sensitive spectrophotometric reagent for PO₄
Catalog # TCIAA5100-1G
Supplier:  TCI
CAS Number:  
undefined
Malachite green oxalate ≥95.0% (by titrimetric analysis) sensitive spectrophotometric reagent for PO₄
Catalog # TCIAA5100-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Green to Dark green powder to crystal
  • Purity (HPLC):
    min. 90.0 area%
  • Purity (Neutralization titration):
    95.0 to 105.0 % (after drying)
  • Lambda max.:
    615.0 to 619.0 nm (H2O)
  • Absorbance (E1%1cm):
    min. 1,800 (H2O, 615.0 to 619.0 nm)

Specification Test Results

About this item

 :
 :927.02 g/mol
Merck Index:13,05722
Melting Pt:∼159 °C
Storage Temperature:Ambient
EINECS:219-441-7
UN:2811
ADR:6.1,III
 :GHS05
 :GHS07
 :GHS08
 :GHS09
CAS Number:2437-29-8
 :18015-76-4
Formula:C₅₂H₅₄N₄O₁₂
MDL Number:MFCD00011766