5-Sulphosalicylic acid dihydrate 99.0-101.0%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:5965-83-3
:
Formula:HO₃SC₆H₃-2-(OH)CO₂H·2H₂O
MDL Number:MFCD00149540
:254.22 g/mol
Merck Index:13,09054
Melting Pt:120 °C
Density:1.695 g/cm³ (20 °C)
Flash Pt:150 °C
Storage Temperature:Ambient
EINECS:202-555-6
UN:2585
ADR:8,III
:GHS07
Specification Test Results
- Meets ACS Reagent Chemical Requirements:99.0 - 101.0%
- For Laboratory,Research,or Manufacturing Use:≤0.02%
- Assay (C₇H₆O₆S2H₂O):99.0 - 101.0 %
- Insoluble Matter:≤ 0.02 %
- Residue after Ignition:≤ 0.10 %
- Chloride (Cl):≤ 0.001 %
- Salicylic Acid (HOC₆H₄COOH):≤ 0.04 %
- Sulfate (SO₄):≤ 0.02 %
- Heavy Metals (as Pb):≤ 0.002 %
- Trace Impurities - Iron (Fe):≤ 0.001 %