Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Cyano-4-methylaniline
3-Cyano-4-methylaniline 97%
3-Cyano-4-methylaniline 97%
Irritant
3-Cyano-4-methylaniline 97%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
3-Cyano-4-methylaniline 97%
Supplier:
Thermo Fisher Scientific
About this item
CAS Number:
50670-64-9
:
Formula:
C₈H₈N₂
MDL Number:
MFCD00017608
:
132.16 g/mol
Melting Pt:
88…91 °C
Storage Temperature:
Ambient
UN:
3439
ADR:
6.1,III
:
GHS07
Related Information
Documentation
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation