Order Entry
Finland
ContactUsLinkComponent
 
 
(1H-Imidazol-4-yl)methanol hydrochloride ≥98.0% (by titrimetric analysis)
  TCIAH0414-1G
 :  TCI
 :  
undefined
 
(1H-Imidazol-4-yl)methanol hydrochloride ≥98.0% (by titrimetric analysis)
  TCIAH0414-1G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance:
    White to Yellow to Orange powder to crystal
  • Purity (Argentmetric Titration):
    min. 98.0 %
  • Melting point:
    107.0 to 112.0 deg-C
  • Solubility in Water:
    very faint turbidity

 

 

 :
 :134.57 g/mol
 :251-150-0
 :GHS07
 :Refrigerator
 :32673-41-9
 :C₄H₇ClN₂O
 :MFCD00012697