1,6-Diphenoxy-2,4-hexadiyne
Catalog # MOLEM79271366
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:262.31 g/mol
CAS Number:30980-37-1
Formula:C₁₈H₁₄O₂
MDL Number:MFCD00041712