Order Entry
Finland
ContactUsLinkComponent
 
 
2-Sulphobenzoic acid monoammonium salt ≥98.0% (by HPLC, titration analysis)
  TCIAS0125-100G
 :  TCI
 :  
undefined
 
2-Sulphobenzoic acid monoammonium salt ≥98.0% (by HPLC, titration analysis)
  TCIAS0125-100G
 :  TCI
 :  

 

  • Pk:
    100 g
  • Seq:
    3401

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 98.0 %
  • Solubility in hot Water:
    almost transparency

 

 

 :
 :230-077-8
 :GHS07
 :6939-89-5
 :C₇H₉NO₅S
 :MFCD00040590