Order Entry
Finland
ContactUsLinkComponent
Isonicotinamidine hydrochloride
Isonicotinamidine hydrochloride
Catalog # MAYBCD05159EA
Supplier:  Thermo Scientific
CAS Number:  
Isonicotinamidine hydrochloride
Catalog # MAYBCD05159EA
Supplier:  Thermo Scientific
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :157.601 g/mol
CAS Number:6345-27-3
Formula:C₆H₇N₃·ClH
MDL Number:MFCD00040385