


3-Isopropoxy-4-(tributylstannyl)-1,2-cyclobutenedione ≥97%
Catalog # H51488.MD
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg
Specifications
About this item
CAS Number:129034-70-4
:
Formula:C₁₉H₃₄O₃Sn
MDL Number:MFCD01319011
:429.17 g/mol
ADR:6.1,III
:GHS06
:GHS08
:GHS07
Storage Temperature:Ambient