
5-Fluoro-8-quinolinol ≥99.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:387-97-3
:
Formula:C₉H₆FNO
MDL Number:MFCD00059754
:163.15 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 99.0 %
- Purity (Nonaqueous Titration):min. 99.0 %
- Melting point:110.0 to 113.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together