N(α)-Acetyl-L-tryptophan ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:1218-34-4
:
Formula:C₁₃H₁₄N₂O₃
MDL Number:MFCD00065976
:246.3 g/mol
EINECS:214-935-9
Specification Test Results
- Appearance:White to Light yellow to Light red powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 96.0 %
- Specific rotation [a]20/D:+25.0 to +27.0 deg (C=1, 1mol/L NaOH sol.)