Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
α,3-Dibromo-4-fluorotoluene
α,3-Dibromo-4-fluorotoluene ≥97%
α,3-Dibromo-4-fluorotoluene ≥97%
α,3-Dibromo-4-fluorotoluene ≥97%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
α,3-Dibromo-4-fluorotoluene ≥97%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
78239-71-1
:
Formula:
C₇H₅Br₂F
MDL Number:
MFCD09037367
:
267.92 g/mol
Melting Pt:
41 °C
Storage Temperature:
Ambient
ADR:
8,III
:
GHS05
Related Information
Documentation
Certificates
Search by Lot Number
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation