Capillary columns, GuardGOLD™
Supplier: Thermo Fisher Scientific
Certificates
About this item
These guard columns extend analytical column lifetime by preventing degradation from harsh 'steam cleaning' water injections.
- Highly deactivated to provide superior inertness, essential for analysis of active compounds
- Protects against column contamination caused by non-volatile materials
Frequently Bought Together