Heating tapes, IT-SiS10
Supplier: ISOPAD
Certificates
About this item
Isopad IT-SiS10 is a preterminated, flexible, and waterproof silicone insulated heating tape. Four silicone insulated heating conductors are embedded in a protective overbraid made of cupro-nickel. The temperature depends on the power and application.
- Maximum operating temperature: 200 °C
- Width×Thickness: 11×5 mm (±10%)
- Maximum power per meter 100 W/m
It is non hazardous and used in ordinary areas. It is used for temperature modification of pipes and structures in industries and laboratories.